2,4-Difluoro-5-nitrophenol
Supplier: Apollo Scientific
Warning
PC8736-1G PC8736-25G PC8736-5G
APOSPC8736-1GEA 21.9 EUR
APOSPC8736-1G APOSPC8736-25G APOSPC8736-5G
2,4-Difluoro-5-nitrophenol
2,4-Difluoro-5-nitrophenol
Formula: C₆H₃F₂NO₃
MW: 175.09 g/mol
Melting Pt: 112…120 °C
Storage Temperature: Ambient
MDL Number: MFCD08443942
CAS Number: 113512-57-5