2,4-Dibromo-6-fluorophenyl isothiocyanate 97%
Supplier: Apollo Scientific
|
Warning
|
PC8745-1G
PC8745-25G
PC8745-5G
APOSPC8745-1GEA
63.8
EUR
APOSPC8745-1G
APOSPC8745-25G
APOSPC8745-5G
2,4-Dibromo-6-fluorophenyl isothiocyanate 97%
2,4-Dibromo-6-fluorophenyl isothiocyanate
Formula:
C₇H₂Br₂FNS Melting Pt: 50…51 °C Storage Temperature: Ambient |
CAS Number:
244022-67-1 UN: 3335 ADR: 9,III |