4'-Chloro-3'-fluoro[1,1'-biphenyl]-2-carbaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 4'-Chloro-3'-fluoro-[1,1'-biphenyl]-2-carboxaldehyde, 4'-Chloro-3'-fluoro-[1,1'-biphenyl]-2-carbaldehyde
PC8780-1G PC8780-500MG PC8780-5G
APOSPC8780-1GEA 306 EUR
APOSPC8780-1G APOSPC8780-500MG APOSPC8780-5G
4'-Chloro-3'-fluoro[1,1'-biphenyl]-2-carbaldehyde
4'-Chloro-3'-fluoro[1,1'-biphenyl]-2-carbaldehyde
Formula: C₁₃H₈ClFO
MW: 234.66 g/mol
Melting Pt: 60…62 °C
Storage Temperature: Ambient
MDL Number: MFCD08443962
CAS Number: 223575-82-4