4,4,4-Trifluoro-3-(trifluoromethyl)crotonic acid 95% + 97%
Supplier: Apollo Scientific
Danger
PC8785-1G PC8785-5G PC8785-250MG
APOSPC8785-1GEA 132 EUR
APOSPC8785-1G APOSPC8785-5G APOSPC8785-250MG
4,4,4-Trifluoro-3-(trifluoromethyl)crotonic acid 95% + 97%
4,4,4-Trifluoro-3-(trifluoromethyl)crotonic acid
Formula: C₅H₂F₆O₂
Boiling Pt: 109…110 °C/120mmHg
Melting Pt: 26…28 °C
Storage Temperature: Ambient
MDL Number: MFCD00013574
CAS Number: 1763-28-6
EINECS: 217-180-3
UN: 3265
ADR: 8,III