5-Chloro-2-fluorobenzaldehyde 98%
Supplier: Apollo Scientific
Warning
PC8801-25G PC8801-100G PC8801-500G
APOSPC8801-25GEA 33.5 EUR
APOSPC8801-25G APOSPC8801-100G APOSPC8801-500G
5-Chloro-2-fluorobenzaldehyde 98%
5-Chloro-2-fluorobenzaldehyde
Formula: C₇H₄ClFO
Melting Pt: 29…32 °C
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD03094518
CAS Number: 96515-79-6