5'-Chloro-2'-fluoroacetophenone 95%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(5-Chloro-2-fluorophenyl)ethanone
PC8803-1G PC8803-5G PC8803-10G
APOSPC8803-1GEA 23.2 EUR
APOSPC8803-1G APOSPC8803-5G APOSPC8803-10G
5'-Chloro-2'-fluoroacetophenone 95%
5'-Chloro-2'-fluoroacetophenone
Formula: C₈H₆ClFO
MW: 172.59 g/mol
Melting Pt: 40…42 °C
Storage Temperature: Ambient
MDL Number: MFCD03094519
CAS Number: 541508-27-4