1-Bromo-5-fluoronaphthalene 97%
Supplier: Apollo Scientific
Warning
PC8838-1G PC8838-25G PC8838-5G
APOSPC8838-1GEA 128 EUR
APOSPC8838-1G APOSPC8838-25G APOSPC8838-5G
1-Bromo-5-fluoronaphthalene 97%
1-Bromo-5-fluoronaphthalene
Formula: C₁₀H₆BrF
MW: 225.0571 g/mol
Boiling Pt: 85…90 °C/0.5mmHg
Melting Pt: 18…19 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD09258916
CAS Number: 315-56-0