3-Cyano-4-fluorobenzaldehyde 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-5-formylbenzonitrile
PC8847-5G PC8847-100G PC8847-25G
APOSPC8847-5GEA 21.9 EUR
APOSPC8847-5G APOSPC8847-100G APOSPC8847-25G
3-Cyano-4-fluorobenzaldehyde 97%
3-Cyano-4-fluorobenzaldehyde
Formula: C₈H₄FNO
MW: 149.1219 g/mol
Melting Pt: 80…84 °C
Storage Temperature: Ambient
MDL Number: MFCD01863558
CAS Number: 218301-22-5
UN: 3439
ADR: 6.1,III