6-Fluoroveratraldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoro-4,5-dimethoxybenzaldehyde, 4,5-Dimethoxy-2-fluorobenzaldehyde, 3,4-Dimethoxy-6-fluorobenzaldehyde
PC8855-1G PC8855-5G
APOSPC8855-1GEA 33.5 EUR
APOSPC8855-1G APOSPC8855-5G
6-Fluoroveratraldehyde
6-Fluoroveratraldehyde
Formula: C₉H₉FO₃
MW: 184.17 g/mol
Melting Pt: 93…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00061108
CAS Number: 71924-62-4