5-Bromo-1,3-dichloro-2-fluorobenzene
Supplier: Apollo Scientific
Warning
Synonyms: 3,5-Dichloro-4-fluorobromobenzene
PC8928-100G PC8928-25G PC8928-5G
APOSPC8928-100GEA 63.8 EUR
APOSPC8928-100G APOSPC8928-25G APOSPC8928-5G
5-Bromo-1,3-dichloro-2-fluorobenzene
5-Bromo-1,3-dichloro-2-fluorobenzene
Formula: C₆H₂BrCl₂F
MW: 243.8900 g/mol
Boiling Pt: 227 °C
Melting Pt: 22.2 °C
Storage Temperature: Ambient
MDL Number: MFCD09996901
CAS Number: 17318-08-0
UN: 3082
ADR: 9+EHS,III