4-Chloro-4'-fluorobutyrophenone

Supplier: Apollo Scientific
Warning

APOSPC8945-100GEA 129 EUR
APOSPC8945-100G
4-Chloro-4'-fluorobutyrophenone
4-Chloro-4'-fluorobutyrophenone
Formula: C₁₀H₁₀ClFO
MW: 200,64 g/mol
Boiling Pt: 120…122 °C/0,7mmHg
Melting Pt: 44352 °C
Density: 1,22 g/cm³ (25 °C)
Flash Pt: >113 °C
Storage Temperature: Ambient
EINECS: 223-395-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR