2-Hydroxy-4-(trifluoromethyl)aniline 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-5-(trifluoromethyl)phenol, 4-Amino-3-hydroxybenzotrifluoride
PC8948-1G PC8948-5G PC8948-10G PC8948-25G
APOSPC8948-1GEA 55.1 EUR
APOSPC8948-1G APOSPC8948-5G APOSPC8948-10G APOSPC8948-25G
2-Hydroxy-4-(trifluoromethyl)aniline 97%
2-Hydroxy-4-(trifluoromethyl)aniline
Formula: C₇H₆F₃NO
MW: 177.1200 g/mol
Melting Pt: 114…115 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08056279
CAS Number: 454-82-0