6-Chloro-8-fluoro[1,2,4]triazolo[4,3-a]pyridine 97%
Supplier: Apollo Scientific
Synonyms: 6-Chloro-8-fluoro-[1,2,4]triazolo[4,3-a]pyridine
PC900613-1G PC900613-25G PC900613-5G
APOSPC900613-1GEA 145 EUR
APOSPC900613-1G APOSPC900613-25G APOSPC900613-5G
6-Chloro-8-fluoro[1,2,4]triazolo[4,3-a]pyridine 97%
6-Chloro-8-fluoro[1,2,4]triazolo[4,3-a]pyridine
Formula: C₆H₃ClFN₃
MW: 171.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09972195
CAS Number: 1020253-21-7