4-Bromo-2-fluoro-5-methylaniline 98%
Supplier: Apollo Scientific
PC900622-100G
PC900622-25G
APOSPC900622-100GEA
376
EUR
APOSPC900622-100G
APOSPC900622-25G
4-Bromo-2-fluoro-5-methylaniline 98%
4-Bromo-2-fluoro-5-methylaniline
Formula:
C₇H₇BrFN MW: 204.0400 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04973744 CAS Number: 418762-26-2 |