5,7-Bis(trifluoromethyl)-3-cyanopyrazolo[1,5-a]pyrimidine 97%
Supplier: Apollo Scientific
Warning
PC9014-1G PC9014-5G
APOSPC9014-1GEA 52.2 EUR
APOSPC9014-1G APOSPC9014-5G
5,7-Bis(trifluoromethyl)-3-cyanopyrazolo[1,5-a]pyrimidine 97%
5,7-Bis(trifluoromethyl)-3-cyanopyrazolo[1,5-a]pyrimidine
Formula: C₉H₂F₆N₄
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00084901
CAS Number: 175276-40-1