N-Ethyl-3-fluorobenzamide 98%

Supplier: Apollo Scientific

PC901491-100G PC901491-25G PC901491-5G
APOSPC901491-100GEA 632 EUR
APOSPC901491-100G APOSPC901491-25G APOSPC901491-5G
N-Ethyl-3-fluorobenzamide 98%
N-Ethyl 3-fluorobenzamide
Formula: C₉H₁₀FNO
Storage Temperature: Ambient
MDL Number: MFCD01338769
CAS Number: 150079-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR