2,6-Bis(trifluoromethyl)-4-hydroxyquinoline 97%
Supplier: Apollo Scientific
Synonyms: 2,6-Bis(trifluoromethyl)quinolin-4-ol
PC9016-1G PC9016-250MG
APOSPC9016-1GEA 126 EUR
APOSPC9016-1G APOSPC9016-250MG
2,6-Bis(trifluoromethyl)-4-hydroxyquinoline 97%
2,6-Bis(trifluoromethyl)-4-hydroxyquinoline
Formula: C₁₁H₅F₆NO
MW: 281.16 g/mol
Melting Pt: 280…290 °C
Storage Temperature: Ambient
MDL Number: MFCD00276581
CAS Number: 35877-04-4