2,8-Bis(trifluoromethyl)quinolin-4-ol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,8-Bis(trifluoromethyl)-4-hydroxyquinoline, 2,8-Bis(trifluoromethyl)-4-quinolinol
PC9017-1G PC9017-25G PC9017-5G
APOSPC9017-1GEA 21.9 EUR
APOSPC9017-1G APOSPC9017-25G APOSPC9017-5G
2,8-Bis(trifluoromethyl)quinolin-4-ol 97%
2,8-Bis(trifluoromethyl)quinolin-4-ol
Formula: C₁₁H₅F₆NO
MW: 281.16 g/mol
Melting Pt: 127…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00075091
CAS Number: 35853-41-9
EINECS: 252-762-0