3,5-Difluoro-N-methylaniline 97%
Supplier: Apollo Scientific
PC901749-100G PC901749-25G
APOSPC901749-100GEA 435 EUR
APOSPC901749-100G APOSPC901749-25G
3,5-Difluoro-N-methylaniline 97%
3,5-Difluoro-N-methylaniline
Formula: C₇H₇F₂N
Storage Temperature: Ambient
MDL Number: MFCD09878315
CAS Number: 470458-51-6