4-(Trifluoromethyl)indole 97%
Supplier: Apollo Scientific
Synonyms: 4-(Trifluoromethyl)-1H-indole
PC902315-1G PC902315-250MG PC902315-5G
APOSPC902315-1GEA 127 EUR
APOSPC902315-1G APOSPC902315-250MG APOSPC902315-5G
4-(Trifluoromethyl)indole 97%
4-(Trifluoromethyl)indole
Formula: C₉H₆F₃N
MW: 185.149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09954766
CAS Number: 128562-95-8