TATU (O-(7-aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-Oxide Tetrafluorobo, TATU, O-(7-Azabenzotriazole-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 2-(3H-[1,2,3]Triazolo[4,5-b]pyridin-3-yl)-1,1,3,3-tetramethyluronium tetrafluoroborate
PC902639-100G PC902639-25G PC902639-5G
APOSPC902639-100GEA 263 EUR
APOSPC902639-100G APOSPC902639-25G APOSPC902639-5G
TATU (O-(7-aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate) 98%
TATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
Formula: C₁₀H₁₅BF₄N₆O
MW: 322.1 g/mol
Melting Pt: 184 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD08064301
CAS Number: 873798-09-5
UN: 1759
ADR: 8 III