2-(2,4-Difluorophenyl)pyridine 98%
Supplier: Apollo Scientific
PC902787-100G PC902787-25G
APOSPC902787-100GEA 409 EUR
APOSPC902787-100G APOSPC902787-25G
2-(2,4-Difluorophenyl)pyridine 98%
2-(2,4-Difluorophenyl)pyridine
Formula: C₁₁H₇F₂N
MW: 191.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07787526
CAS Number: 391604-55-0