2-Chloro-4,5-difluorobenzaldehyde 95%
Supplier: Apollo Scientific
PC904911-1G PC904911-5G
APOSPC904911-1GEA 26.1 EUR
APOSPC904911-1G APOSPC904911-5G
2-Chloro-4,5-difluorobenzaldehyde 95%
2-Chloro-4,5-difluorobenzaldehyde
Formula: C₇H₃ClF₂O
MW: 176.5500 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07782050
CAS Number: 165047-23-4