3-(1,1,2,2-Tetrafluoroethoxy)benzoic acid
Supplier: Apollo Scientific
Warning
PC9058-100G PC9058-25G
APOSPC9058-100GEA 0 EUR
APOSPC9058-100G APOSPC9058-25G
3-(1,1,2,2-Tetrafluoroethoxy)benzoic acid
3-(1,1,2,2-Tetrafluoroethoxy)benzoic acid
Formula: C₉H₆F₄O₃
Melting Pt: 127…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00155925
CAS Number: 70126-48-6