2-Bromo-4-fluoro-6-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
Synonyms: 6-Bromo-α,α,α,4-tetrafluoro-o-toluidine
PC906319-25G PC906319-5G PC906319-100G
APOSPC906319-25GEA 132 EUR
APOSPC906319-25G APOSPC906319-5G APOSPC906319-100G
2-Bromo-4-fluoro-6-(trifluoromethyl)aniline 98%
2-Bromo-4-fluoro-6-(trifluoromethyl)aniline
Formula: C₇H₄BrF₄N
MW: 258.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973756
CAS Number: 875664-27-0