2-Chloro-5-fluoro-3-methylbenzoic acid 97%
Supplier: Apollo Scientific
PC906340-1G PC906340-5G
APOSPC906340-1GEA 219 EUR
APOSPC906340-1G APOSPC906340-5G
2-Chloro-5-fluoro-3-methylbenzoic acid 97%
2-Chloro-5-fluoro-3-methylbenzoic acid
Formula: C₈H₆ClFO
MW: 188.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23712098
CAS Number: 1427416-28-1