3-Fluoro-N-methylaniline 97%
Supplier: Apollo Scientific
PC906345-25G PC906345-5G
APOSPC906345-25GEA 242 EUR
APOSPC906345-25G APOSPC906345-5G
3-Fluoro-N-methylaniline 97%
3-Fluoro-N-methylaniline
Formula: C₇H₈FN
Storage Temperature: Ambient
MDL Number: MFCD02683095
CAS Number: 1978-37-6