2-Chloro-4-fluoro-3-methylbenzoic acid 98%
Supplier: Apollo Scientific
PC906389-250MG
APOSPC906389-250MGEA 68.3 EUR
APOSPC906389-250MG
2-Chloro-4-fluoro-3-methylbenzoic acid 98%
2-Chloro-4-fluoro-3-methylbenzoic acid
Formula: C₈H₆ClFO₂
MW: 188.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846444
CAS Number: 173315-54-3