2,3,5,6-Tetrafluoropyridine 98%
Supplier: Apollo Scientific
Warning
PC9064-25G PC9064-5G PC9064-100G PC9064-10G
APOSPC9064-25GEA 134 EUR
APOSPC9064-25G APOSPC9064-5G APOSPC9064-100G APOSPC9064-10G
2,3,5,6-Tetrafluoropyridine 98%
2,3,5,6-Tetrafluoropyridine
Formula: C₅HF₄N
MW: 151.06 g/mol
Boiling Pt: 102 °C
Density: 1.499 g/cm³ (25 °C)
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00011734
CAS Number: 2875-18-5
UN: 1993
ADR: 3,III