(2-(Trifluoromethoxy)phenyl)acetic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(2-(Trifluoromethoxy)phenyl)acetic acid
PC9067-1G PC9067-5G PC9067-10G
APOSPC9067-1GEA 21.9 EUR
APOSPC9067-1G APOSPC9067-5G APOSPC9067-10G
(2-(Trifluoromethoxy)phenyl)acetic acid 98%
(2-(Trifluoromethoxy)phenyl)acetic acid
Formula: F₃COC₆H₄CH₂COOH
MW: 220.15 g/mol
Melting Pt: 54…56 °C
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00236325
CAS Number: 220239-67-8