3'-Amino-4'-fluoroacetophenone 95%

Supplier: Apollo Scientific

PC906736-5G
APOSPC906736-5GEA 105 EUR
APOSPC906736-5G
3'-Amino-4'-fluoroacetophenone 95%
3'-Amino-4'-fluoroacetophenone
Formula: C₈H₈FNO
MW: 153.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00115400
CAS Number: 2002-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR