2-Bromo-4-(trifluoromethyl)pyrimidine 97%
Supplier: Apollo Scientific
Warning
PC908649-1G PC908649-250MG PC908649-5G
APOSPC908649-1GEA 74.1 EUR
APOSPC908649-1G APOSPC908649-250MG APOSPC908649-5G
2-Bromo-4-(trifluoromethyl)pyrimidine 97%
2-Bromo-4-(trifluoromethyl)pyrimidine
Formula: C₅H₂BrF₃N₂
Boiling Pt: 55…56 °C
Melting Pt: 21…23 °C
Density: 1.826 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD09802248
CAS Number: 785777-87-9