6,7-Difluoro-3,4-dihydronaphthalen-1(2H)-one 97%
Supplier: Apollo Scientific
PC910926-100MG PC910926-1G PC910926-250MG
APOSPC910926-100MGEA 58 EUR
APOSPC910926-100MG APOSPC910926-1G APOSPC910926-250MG
6,7-Difluoro-3,4-dihydronaphthalen-1(2H)-one 97%
6,7-Difluoro-3,4-dihydronaphthalen-1(2H)-one
Formula: C₁₀H₈F₂O
MW: 182.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08234366
CAS Number: 137114-68-2