2,3-Dibromo-1,1,1,4,4,4-hexafluorobutane 97%
Supplier: Apollo Scientific
Warning
PC9125-1G PC9125-5G
APOSPC9125-1GEA 69.6 EUR
APOSPC9125-1G APOSPC9125-5G
2,3-Dibromo-1,1,1,4,4,4-hexafluorobutane 97%
2,3-Dibromo-1,1,1,4,4,4-hexafluorobutane
Formula: C₄H₂Br₂F₆
Boiling Pt: 62 °C/300mmHg
Density: 1.8 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 384-50-9