Potassium(4-fluorophenyl)trifluoroborate
Supplier: Apollo Scientific
Warning
Synonyms: (4-Fluorophenyl)trifluoroboric acid potassium salt, Potassium trifluoro(4-fluorophenyl)borate
PC9162-1G PC9162-5G PC9162-25G
APOSPC9162-1GEA 21.9 EUR
APOSPC9162-1G APOSPC9162-5G APOSPC9162-25G
Potassium(4-fluorophenyl)trifluoroborate
Potassium (4-fluorophenyl)trifluoroborate
Formula: C₆H₄BF₄K
Melting Pt: 297 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01318170
CAS Number: 192863-35-7