2-Fluoro-4-methoxybenzoyl chloride 95%
Supplier: Apollo Scientific
PC9192-5G PC9192-1G
APOSPC9192-5GEA 357 EUR
APOSPC9192-5G APOSPC9192-1G
2-Fluoro-4-methoxybenzoyl chloride 95%
2-Fluoro-4-methoxybenzoyl chloride
Formula: C₈H₆ClFO₂
Melting Pt: 40…42 °C
Storage Temperature: Ambient
MDL Number: MFCD08146649
CAS Number: 321-24-4