4-(Trifluoromethyl)anthranilic acid 98%
Supplier: Apollo Scientific
Warning
PC9193-25G PC9193-5G PC9193-100G
APOSPC9193-25GEA 66.7 EUR
APOSPC9193-25G APOSPC9193-5G APOSPC9193-100G
4-(Trifluoromethyl)anthranilic acid 98%
4-(Trifluoromethyl)anthranilic acid
Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
Melting Pt: 175…176 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00519334
CAS Number: 402-13-1