2-Bromo-4'-methoxy-3'-(trifluoromethyl)acetophenone 97%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Methoxy-3-(trifluoromethyl)phenacyl bromide
PC9207-1G PC9207-250MG PC9207-5G
APOSPC9207-1GEA 241 EUR
APOSPC9207-1G APOSPC9207-250MG APOSPC9207-5G
2-Bromo-4'-methoxy-3'-(trifluoromethyl)acetophenone 97%
2-Bromo-4'-methoxy-3'-(trifluoromethyl)acetophenone
Formula: C₁₀H₈BrF₃O₂
Melting Pt: 71…72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094546
CAS Number: 541550-00-9
UN: 3261
ADR: 8 III