2-Chloro-5-(trifluoromethyl)nicotinic acid
Supplier: Apollo Scientific
Warning
PC9219-1G PC9219-250MG PC9219-5G
APOSPC9219-1GEA 26.1 EUR
APOSPC9219-1G APOSPC9219-250MG APOSPC9219-5G
2-Chloro-5-(trifluoromethyl)nicotinic acid
2-Chloro-5-(trifluoromethyl)nicotinic acid
Formula: C₇H₃ClF₃NO₂
MW: 225.55 g/mol
Melting Pt: 173…178 °C
Storage Temperature: Ambient
MDL Number: MFCD08741353
CAS Number: 505084-59-3
UN: 2811
ADR: 6.1 III