2-Chloro-3-(trifluoromethyl)quinoline 95%

Supplier: Apollo Scientific

PC9221-1G PC9221-500MG PC9221-5G
APOSPC9221-1GEA 234 EUR
APOSPC9221-1G APOSPC9221-500MG APOSPC9221-5G
2-Chloro-3-(trifluoromethyl)quinoline 95%
2-Chloro-3-(trifluoromethyl)quinoline
Formula: C₁₀H₅ClF₃N
MW: 231.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08741354
CAS Number: 25199-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR