2-Fluoro-6-iodobenzaldehyde 97%
Supplier: Apollo Scientific
Danger
Synonyms: 6-Fluoro-2-Iodobenzaldehyde
PC9246-1G PC9246-5G PC9246-25G
APOSPC9246-1GEA 21.9 EUR
APOSPC9246-1G APOSPC9246-5G APOSPC9246-25G
2-Fluoro-6-iodobenzaldehyde 97%
2-Fluoro-6-iodobenzaldehyde
Formula: C₇H₄FIO
MW: 250.01 g/mol
Melting Pt: 36…40 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD07371506
CAS Number: 146137-72-6
UN: 2811
ADR: 6.1 III