2-Bromo-5-chlorobenzotrifluoride 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-5-chlorobenzotrifluoride, 1-Bromo-4-chloro-2-(trifluoromethyl)benzene
PC9265-25G PC9265-5G PC9265-100G
APOSPC9265-25GEA 24.8 EUR
APOSPC9265-25G APOSPC9265-5G APOSPC9265-100G
2-Bromo-5-chlorobenzotrifluoride 99%
2-Bromo-5-chlorobenzotrifluoride
Formula: C₇H₃BrClF₃
MW: 259.45 g/mol
Boiling Pt: 179…180 °C
Melting Pt: 18…20 °C
Density: 1.759 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00010308
CAS Number: 344-65-0