6-Fluoro-o-toluonitrile 99%
Supplier: Apollo Scientific
Danger
PC9287-1G PC9287-5G PC9287-10G
APOSPC9287-1GEA 21.9 EUR
APOSPC9287-1G APOSPC9287-5G APOSPC9287-10G
6-Fluoro-o-toluonitrile 99%
6-Fluoro-o-toluonitrile
Formula: C₈H₆FN
Melting Pt: 43…45 °C
Flash Pt: 90.5 °C
Storage Temperature: Ambient
MDL Number: MFCD06660277
CAS Number: 198633-76-0