Ethyl-2-fluoropropionate 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Fluoropropionic Acid Ethyl Ester, Ethyl 2-fluoropropionate
PC9352-100G PC9352-25G PC9352-5G
APOSPC9352-100GEA 101 EUR
APOSPC9352-100G APOSPC9352-25G APOSPC9352-5G
Ethyl-2-fluoropropionate 97%
Ethyl-2-fluoropropionate
Formula: C₅H₉FO₂
MW: 120.12 g/mol
Boiling Pt: 121…125 °C
Density: 1.03 g/cm³ (25 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD01320801
CAS Number: 349-43-9
UN: 3272
ADR: 3 III