4-(4-Fluorophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile 97%
Supplier: Apollo Scientific
Warning
PC9360-1G PC9360-5G
APOSPC9360-1GEA 227 EUR
APOSPC9360-1G APOSPC9360-5G
4-(4-Fluorophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile 97%
4-(4-Fluorophenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile
Formula: C₁₁H₆FNO₂
Melting Pt: 210…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00139244
CAS Number: 50691-04-8