Methyl-2-amino-6-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4,5,6,7-tetrahydrothieno[2,3-c]pyridine 97%
Supplier: Apollo Scientific
Warning
PC9389-1G PC9389-500MG
APOSPC9389-1GEA 604 EUR
APOSPC9389-1G APOSPC9389-500MG
Methyl-2-amino-6-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4,5,6,7-tetrahydrothieno[2,3-c]pyridine 97%
Methyl-2-amino-6-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4,5,6,7-tetrahydrothieno[2,3-c]pyridine
Formula: C₁₅H₁₃ClF₃N₃O₂S
MW: 391.8 g/mol
Melting Pt: 149…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00180791
CAS Number: 321430-29-9