Methyl-3-(2-fluoro-4-nitrophenoxy)-2-thiophenecarboxylate 97%
Supplier: Apollo Scientific
Warning
PC9409-1G PC9409-100MG
APOSPC9409-1GEA 56.7 EUR
APOSPC9409-1G APOSPC9409-100MG
Methyl-3-(2-fluoro-4-nitrophenoxy)-2-thiophenecarboxylate 97%
Methyl-3-(2-fluoro-4-nitrophenoxy)-2-thiophenecarboxylate
Formula: C₁₂H₈FNO₅S
MW: 297.26 g/mol
Melting Pt: 112…114 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00202715
CAS Number: 252026-46-3