1-Methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde 97%
Supplier: Apollo Scientific
PC9433-1G PC9433-250MG
APOSPC9433-1GEA 220 EUR
APOSPC9433-1G APOSPC9433-250MG
1-Methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde 97%
1-Methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
Formula: C₁₂H₉F₃N₂O₂
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD01936001
CAS Number: 109925-42-0