1-Methyl-3-(trifluoromethyl)-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carbaldehyde 97%
Supplier: Apollo Scientific
PC9461-1G PC9461-500MG PC9461-5G
APOSPC9461-1GEA 533 EUR
APOSPC9461-1G APOSPC9461-500MG APOSPC9461-5G
1-Methyl-3-(trifluoromethyl)-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carbaldehyde 97%
1-Methyl-3-(trifluoromethyl)-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carbaldehyde
Formula: C₁₃H₈F₆N₂O₂
MW: 338.21 g/mol
Melting Pt: 81…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00243685
CAS Number: 158712-22-2