4-Oxo-3-[3-(trifluoromethyl)phenyl]3,4-dihydro-1-phthalazinecarboxylic acid 97%
Supplier: Apollo Scientific
PC9479-1G PC9479-500MG PC9479-5G
APOSPC9479-1GEA 275 EUR
APOSPC9479-1G APOSPC9479-500MG APOSPC9479-5G
4-Oxo-3-[3-(trifluoromethyl)phenyl]3,4-dihydro-1-phthalazinecarboxylic acid 97%
4-Oxo-3-[3-(trifluoromethyl)phenyl]3,4-dihydro-1-phthalazinecarboxylic acid
Formula: C₁₆H₉F₃N₂O₃
Melting Pt: 210…213 °C
Storage Temperature: Ambient
MDL Number: MFCD00138613
CAS Number: 339021-26-0